missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Equine IgM Heavy Chain Secondary Antibody, DyLight 755, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7423IR
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Equine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Horse IgM | |
| 1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction