Learn More
FOXQ1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FOXQ1.
Brand: Abnova PAB6444.100ug
Description
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis (Bieller et al., 2001 [PubMed 11747606]).[supplied by OMIM
Sequence: KLEVFVPRAAHGD-CSpecifications
| FOXQ1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| FOXQ1 | |
| HFH1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 94234 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FOXQ1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_150285 | |
| FOXQ1 | |
| A synthetic peptide corresponding to human FOXQ1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only