missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabpb2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Gabpb2.
Brand: Abnova PAB7287.100ug
This item is not returnable.
View return policy
Description
Sequence: C-KSGRTTEPHTNSpecifications
| Gabpb2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
| Gabpb2 | |
| 1810015F01Rik/5830427M07/9430006E19Rik/A430024B14Rik/AV050852/Gabpb2-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 213054 | |
| Mouse | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of Gabpb2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_766100.1;NP_084161.1 | |
| Gabpb2 | |
| A synthetic peptide corresponding to mouse Gabpb2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_766100.1, NP_084161.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction