missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against full-length recombinant GFP.
Brand: Abnova PAB10341.1mg
This item is not returnable.
View return policy
Specifications
| GFP | |
| Polyclonal | |
| Goat polyclonal antibody raised against full length recombinant GFP. | |
| In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Recombinant protein corresponding to full length jellyfish Aequorea victoria GFP. | |
| RUO | |
| No reaction was observed against human, Mouse or Rat serum proteins. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:40000) Western Blot (1:400-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| Goat | |
| 1 mg | |
| Primary | |
| Jellyfish | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction