missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucoamylase 1 (Rhizopus), Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native Glucoamylase 1 (Rhizopus).
Brand: Abnova PAB28051.100ug
This item is not returnable.
View return policy
Specifications
| Glucoamylase 1 (Rhizopus) | |
| Polyclonal | |
| Goat polyclonal antibody raised against native Glucoamylase 1 (Rhizopus). | |
| Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Native purified Rhizopus Glucoamylase 1. | |
| RUO | |
| Fungi | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:5000 - 1:25000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| Goat | |
| 100 μg | |
| Primary | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction