missing translation for 'onlineSavingsMsg'
Learn More

GULP1, Goat, Polyclonal Antibody, Abnova™

Product Code. 16019467
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16019467 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16019467 Supplier Abnova Supplier No. PAB6174.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat polyclonal antibody raised against synthetic peptide of GULP1.

The prompt clearance of cells undergoing apoptosis is critical during embryonic development, normal tissue turnover, inflammation, and autoimmunity. CED6 is an evolutionarily conserved adaptor protein required for efficient engulfment of apoptotic cells by phagocytes.[supplied by OMIM

Sequence: EGTVFCLDPLDSRC

Specifications

Antigen GULP1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of GULP1.
Dilution ELISA (1:64000) Western blot (0.2-0.6 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene GULP1
Gene Accession No. NP_057399.1
Gene Alias CED-6/CED6/FLJ31156/GULP
Gene Symbols GULP1
Host Species Goat
Immunogen A synthetic peptide corresponding to human GULP1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51454
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.