Learn More
HAMP, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of HAMP.
Brand: Abnova PAB15632.100ug
Description
The product encoded by this gene is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity. Mutations in this gene cause hemochromatosis type 2B, also known as juvenile hemochromatosis, a disease caused by severe iron overload that results in cardiomyopathy, cirrhosis, and endocrine failure. [provided by RefSeq
Sequence: TSGSVFPQQTGQLAE-CSpecifications
| HAMP | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| HAMP | |
| HAMP | |
| A synthetic peptide corresponding to human HAMP. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of HAMP. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| HEPC/HEPCIDIN/HFE2B/LEAP-1/LEAP1/PLTR | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 57817 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only