missing translation for 'onlineSavingsMsg'
Learn More

hepatitis C virus Goat anti-Viruses, Polyclonal Antibody, Abnova™

Product Code. 16081017
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16081017 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16081017 Supplier Abnova Supplier No. PAB13974.1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat polyclonal antibody raised against partial recombinant hepatitis C virus.

Specifications

Antigen hepatitis C virus
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against partial recombinant hepatitis C virus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Recombinant protein corresponding to NS-4 region of hepatitis C virus (genotype 1a).
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Hepatitis C Virus, NS-4 Region.
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.