missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Horse IgG F(ab')2 fragment, HRP, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10651.2mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Horse IgG F(ab')2 fragment | |
| Polyclonal | |
| Goat anti-horse IgG recognizes horse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicin sulfate) | |
| Horse IgG F(ab')2 fragment | |
| 2 mg | |
| Equine | |
| Lyophilized |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction