Learn More
HOXA9 Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HOXA9.
Brand: Abnova PAB18560.100ug
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. This gene is highly similar to the abdominal-B (Abd-B) gene of Drosophila. A specific translocation event which causes a fusion between this gene and the NUP98 gene has been associated with myeloid leukemogenesis. [provided by RefSeq
Sequence: C-QPPRQAATLAEHPDSpecifications
| HOXA9 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| HOXA9 | |
| HOXA9 | |
| A synthetic peptide corresponding to amino acids 37-50 at internal region of human HOXA9. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of HOXA9. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| ABD-B/HOX1/HOX1.7/HOX1G/MGC1934 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3205 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only