Learn More
HOXB13, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HOXB13.
Brand: Abnova PAB7144.100ug
Description
This gene encodes a transcription factor that belongs to the homeobox gene family. Genes of this family are highly conserved among vertebrates and essential for vertebrate embryonic development. This gene has been implicated to play a role in fetal skin development and cutaneous regeneration. In mice, a similar gene was shown to exhibit temporal and spatial colinearity in the main body axis of the embryo, but was not expressed in the secondary axes, which suggests functions in body patterning along the axis. This gene and other HOXB genes form a gene cluster at chromosome the 17q21-22 region. [provided by RefSeq
Sequence: C-KKVLAKVKNSATPSpecifications
| HOXB13 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| HOXB13 | |
| HOXB13 | |
| A synthetic peptide corresponding to human HOXB13. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of HOXB13. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PSGD | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10481 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only