Learn More
HPS6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HPS6.
Brand: Abnova PAB6733.100ug
Description
This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 5 protein. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 6. [provided by RefSeq
Sequence: LRTELRDYRGLECSpecifications
| HPS6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| HPS6 | |
| HPS6 | |
| A synthetic peptide corresponding to human HPS6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of HPS6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FLJ22501/MGC20522/RP11-302K17.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 79803 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only