missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Albumin Polyclonal Antibody, MP Biomedicals
Brand: MP Biomedicals 0855235
This item is not returnable.
View return policy
Description
Host Species: Goat
Species Reactivity: Human
Specifications
| Albumin | |
| HRP | |
| 2 mL | |
| Human |
| Polyclonal | |
| Goat | |
| Primary | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction