missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA/IgG/IgM (H+L) Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72757H
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| PBS | |
| Human IgG, IgA and IgM whole molecules | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| HRP | |
| Goat | |
| Multi-step | |
| RUO | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction