missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9295.1mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Human IgG F(ab')2 fragment | |
| Polyclonal | |
| Goat anti-human IgG recognizes human IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Human IgG F(ab')2 fragment | |
| 1 mg | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. | |
| Secondary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction