Learn More
Invitrogen™ Goat anti-Human IgG Fc Secondary Antibody, Biotin, eBioscience™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ 13-4998-83
Description
Description: This goat antibody reacts with the Fc fragment of heavy chains on human IgG but notwith the light chains on most human immunoglobulins. No antibody was detected against human IgM or IgA, or against nonimmunoglobulinserum proteins. This antibody has been tested to ensure minimalcross-reaction with bovine, horse, and mouse serum proteins, but the antibody may cross-react with immunoglobulins fromother species. Applications Reported: Biotin goat anti-Human IgG (Fcg Frag Specific) has been reported for use in flow cytometric analysis. Applications Tested: Biotin goat anti-Human IgG (Fc gamma Frag Specific) has been tested by flow cytometric analysis to detect purified human Fc-tagged proteins. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. We recommend a starting concentration of 0.03 μg of Streptavidin PE (Product # 12-4317-87) for detection of Biotin Anti-Human IgG Fcg Fragment Specific. Filtration: 0.2 μm post-manufacturing filtered.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgG Fc | |
| Polyclonal | |
| Biotin | |
| Goat | |
| 200 μg | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Affinity chromatography | |
| RUO | |
| Human | |
| Secondary Antibody | |
| Ig |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.