missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7446V
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Electron Microscopy
Specifications
| IgG Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Human IgG-Fc Fragment | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Electron Microscopy | |
| DyLight 405 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction