missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60678
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot.
Specifications
| IgG Fc | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Human IgG F(c) fragment | |
| 1 mg | |
| This antibody was pre-adsorbed against Mouse Serum Proteins. No reaction was observed against Human IgG F(ab) or Mouse Serum Proteins. Specificity was confirmed by ELISA minimal cross reactivity against Mouse IgG. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| 1 mg/mL | |
| Western Blot 1:1000-1:5000, ELISA 1:15000-1:1:60000, Immunohistochemistry 1:1000-1:3000, Dot Blot | |
| Goat | |
| Affinity Purified | |
| Secondary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction