missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
€ 107.00
Specifications
| Antigen | IgG Fc |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.Specifications
| IgG Fc | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| O75015, O75015, O75015 | |
| IgG | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy gamma chains on human IgG. This antibody has been pre-adsorbed against bovine, horse, mouse, and rabbit serum proteins. |
| 4.5 mg/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| Purified Human IgG, Fc fragment | |
| Secondary | |
| Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title