missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc, γ-Specific Polyclonal Secondary Antibody, AP, MP Biomedicals
Brand: MP Biomedicals 0859289
This item is not returnable.
View return policy
Description
Host Species: Goat
Species Reactivity: Human
Specifications
| IgG Fc, gamma-specific | |
| Alkaline Phosphatase (AP) | |
| 1 mL | |
| Human |
| Polyclonal | |
| Goat | |
| Secondary | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction