missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, AP

Product Code. 10363143
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10363143 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10363143 Supplier Invitrogen™ Supplier No. 628422
  • ONLINE EXCLUSIVE PRICE
    € 235.00
    € 188.00 / 1mL
    Save € 47.00
    20% Off
View all promotions
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Secondary Antibody

Goat anti-Human IgG (H+L) Secondary Antibody for ELISA, IHC (P), IP, Western Blot

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (H+L)
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation 0.05M tris with 1mM MgCl2, 40% glycerol, 1% BSA, 0.15M NaCl and 0.1% sodium azide; pH 7.5
Host Species Goat
Immunogen Gamma Immunoglobin.
Purification Method Affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.