missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (Light chain) Superclonal™ Secondary Antibody

Goat Recombinant Superclonal Secondary Antibody

€ 119.00 - € 287.00

Specifications

Antigen Human IgG (Light chain)
Concentration 1.0 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17836992
View Documents
Invitrogen™
A56853
250 μL
€ 287.00
250µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17885782
View Documents
Invitrogen™
A5685350UL
50 μL
€ 119.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Goat anti-Human IgG (Light Chain Kappa+Lambda), Superclonal™ Recombinant Secondary Antibody specifically detects both Kappa and the Lambda light chains of human IgG, with no cross-reactivity to heavy chains and other species immunoglobulins. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. Superclonal™ Recombinant antibodies are a defined pool of recombinant monoclonal antibodies, manufactured by selecting and pooling individual clones to provide complementary, specific, and optimal epitope coverage to achieve enhanced performance. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Human IgG (Light chain)
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Recombinant Superclonal
Liquid
IgG
Human
Purified Human IgG.
Secondary Antibody
1.0 mg/mL
ELISA, Western Blot, Immunocytochemistry
Unconjugated
Goat
RUO
PBS with 0.09% sodium azide; pH 7.4
Secondary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.