missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgM Fc Secondary Antibody, DyLight 488, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7436G
This item is not returnable.
View return policy
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgM Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Human IgM | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction