Learn More
Goat anti-Human IgM Fc Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NB7438
Product Code. 18770374
  • € 295.00 / 1mL
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin
Affinity Purified