missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human kappa light chain,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9278.1mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Human kappa light chain | |
| Polyclonal | |
| Goat anti-human IgG recognizes human kappa light chain. This secondary antibody was purified using antigen affinity chromatography. | |
| In 100mM Na2B4O7, 75mM NaCl, 5mM EDTA, pH 8.0 (0.01% sodium azide) | |
| Human kappa light chain | |
| 1 mg | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction