missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, DyLight 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7463UV
This item is not returnable.
View return policy
Description
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Kappa Light Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Human Kappa Light Chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction