missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, DyLight 680, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75060FR
This item is not returnable.
View return policy
Description
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Specifications
| Kappa Light Chain | |
| Polyclonal | |
| Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Purified Human Kappa light chain | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction