missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Lambda Light Chain Secondary Antibody, PerCP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7468PCP
This item is not returnable.
View return policy
Description
Lambda Light Chain Polyclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Lambda Light Chain | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PerCP | |
| IGLC, immunoglobulin lambda constant group | |
| Human Lambda Light Chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction