Learn More
IGF2BP2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of IGF2BP2.
Brand: Abnova PAB7566.100ug
Description
This gene encodes a member of the IGF-II mRNA-binding protein (IMP) family. The protein encoded by this gene contains several four KH domains and two RRM domains. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: C-KQQEQKYPQGVASQRSpecifications
| IGF2BP2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| IGF2BP2 | |
| IGF2BP2 | |
| A synthetic peptide corresponding to human IGF2BP2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_006539.3 and NP_001007226.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of IGF2BP2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| IMP-2/IMP2/VICKZ2/p62 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10644 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only