missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL7R, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Il7r.
Brand: Abnova PAB19724.100ug
This item is not returnable.
View return policy
Description
Sequence: DDHSFWCHSQLESpecifications
| IL7R | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of Il7r. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| CD127/IL-7Ralpha/MGC107557 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 16197 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| Il7r | |
| Il7r | |
| A synthetic peptide corresponding to N-terminus of mouse Il7r. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction