missing translation for 'onlineSavingsMsg'
Learn More

IRF5, Goat, Polyclonal Antibody, Abnova™

Product Code. 16059507
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16059507 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16059507 Supplier Abnova Supplier No. PAB6227.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat polyclonal antibody raised against synthetic peptide of IRF5.

This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist. [provided by RefSeq

Sequence: C-QGPWPMHPAGMQ

Specifications

Antigen IRF5
Applications ChIP Assay, ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of IRF5.
Dilution ELISA (1:32000) Western blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene IRF5
Gene Alias SLEB10
Gene Symbols IRF5
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human IRF5.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3663
Test Specificity This antibody is expected to recognize all four reported isoforms (NP_002191.1, NP_1160.32.1, NP_001092097.1, NP_001092099.1). Reported variants represent identical protein: NP_001092101.1, NP_001092098.1, NP_001092097.1 Reported variants represent identical protein: NP_1160.32.1, NP_001092100.1
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.