missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITM2B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of ITM2B.
Brand: Abnova PAB15636.100ug
This item is not returnable.
View return policy
Description
Sequence: C-HDKETYKLQRRETIKSpecifications
| ITM2B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ITM2B | |
| ITM2B | |
| A synthetic peptide corresponding to amino acids at internal region of human ITM2B. | |
| 100 μg | |
| Primary | |
| Approximately 33 KDa band observed in human brain (cerebellum and hippocampus) lysates (calculated MW of 30.3 KDa according to NP_068839.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of ITM2B. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| ABRI/BRI/BRI2/BRICD2B/E25B/E3-16/FBD | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9445 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction