Learn More
KCNQ5, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of KCNQ5.
Brand: Abnova PAB15641.100ug
Description
This gene is a member of the KCNQ potassium channel gene family that is differentially expressed in subregions of the brain and in skeletal muscle. The protein encoded by this gene yields currents that activate slowly with depolarization and can form heteromeric channels with the protein encoded by the KCNQ3 gene. Currents expressed from this protein have voltage dependences and inhibitor sensitivities in common with M-currents. They are also inhibited by M1 muscarinic receptor activation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-ESTDALSLPHVKLKSpecifications
| KCNQ5 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) The optimal working dilution should be determined by the end user. | |
| KCNQ5 | |
| KCNQ5 | |
| A synthetic peptide corresponding to human KCNQ5. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of KCNQ5. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| Kv7.5 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 56479 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only