missing translation for 'onlineSavingsMsg'
Learn More

LDHC Goat anti-Mouse, Polyclonal Antibody, Abnova™

Product Code. 16000588
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16000588 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16000588 Supplier Abnova Supplier No. PAB7509.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat polyclonal antibody raised against synthetic peptide of LDHC.

Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region. [provided by RefSeq]

Sequence: C-DSDKEHWKNVHKQ

Specifications

Antigen LDHC
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of LDHC.
Dilution ELISA (1:16000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene LDHC
Gene Alias CT32/LDH3/LDHX/MGC111073
Gene Symbols LDHC
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 221-233 of human LDHC.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3948
Test Specificity Both variants represent identical product (NP_002292.1 and NP_059144.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.