Learn More
MAN2A1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MAN2A1.
Brand: Abnova PAB7421.100ug
Description
This gene encodes a protein which is a member of family 38 of the glycosyl hydrolases. The protein is located in the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus. [provided by RefSeq
Sequence: C-KISSDIKSQNRSpecifications
| MAN2A1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| MAN2A1 | |
| MAN2A1 | |
| A synthetic peptide corresponding to internal region of human MAN2A1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MAN2A1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| GOLIM7/MANA2/MANII | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4124 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only