Learn More
MGAT1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MGAT1.
Brand: Abnova PAB18956.100ug
Description
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. UDP-N-acetylglucosamine:alpha-3-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I is a medial-Golgi enzyme essential for the synthesis of hybrid and complex N-glycans. The protein, encoded by a single exon, shows typical features of a type II transmembrane protein. The protein is believed to be essential for normal embryogenesis. Several variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: C-QVEKVRTNDRSpecifications
| MGAT1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MGAT1 | |
| MGAT1 | |
| A synthetic peptide corresponding to amino acids at internal region of human MGAT1. | |
| 100 μg | |
| Primary | |
| Reported variants represent identical protein: NP_002397.2, NP_001108090.1, NP_001108091.1, NP_001108092.1, NP_001108089.1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MGAT1. | |
| In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| GLCNAC-TI/GLCT1/GLYT1/GNT-1/GNT-I/MGAT | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4245 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only