Learn More
Goat anti-Monkey IgM Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-73726
Product Code. 18781494
  • € 477.00 / 1mg
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA, Immunohistochemistry
LYOPH
Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Western Blot 1:1000 - 1:5000, ELISA 1:65000 - 1:160000, Immunohistochemistry 1:500 -1:2500
Affinity Purified
IgM