missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Monkey IgM Heavy Chain Secondary Antibody, Texas Red, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73560
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| Texas Red | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
| LYOPH | |
| Western Blot, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| IGHM | |
| Monkey IgM u (mu) heavy chain | |
| 1 mg | |
| Secondary | |
| No reaction was observed against other monkey heavy or light chain proteins. | |
| Monkey | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering