Learn More
Invitrogen™ Goat anti-Mouse IgA Cross-Adsorbed Secondary Antibody, HRP
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA174397
This item is not returnable.
View return policy
Description
This antibody recognizes Mouse and Rat IgA and has been cross absorbed against Mouse IgM, IgG1, IgG2a, IgG2b and IgG3. This antibody will not react with Human samples.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgA Cross-Adsorbed | |
| Polyclonal | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.4 | |
| Mouse IgA paraproteins. | |
| 1 mL | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| HRP | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |