missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgA/IgG/IgM (H+L) Secondary Antibody, Allophycocyanin/Cy7, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72780APCCY7
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| PBS | |
| Mouse IgG IgA and IgM whole molecule | |
| 500 μg | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry | |
| APC-Cyanine7 | |
| Goat | |
| Multi-step | |
| RUO | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction