missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgA Secondary Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68523R
This item is not returnable.
View return policy
Description
IgA Polyclonal antibody specifically detects IgA in Mouse samples. It is validated for ELISA
Specifications
| IgA | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
| ELISA | |
| DyLight 550 | |
| Immunoglobulin A | |
| Mouse IgA paraproteins | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| 3493 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction