Learn More
Invitrogen™ Goat anti-Mouse IgE Secondary Antibody
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA129273
This item is not returnable.
View return policy
Description
PA1-29273 detects Mouse IgE from mouse samples.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgE | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Highly purified mouse IgE hybridoma protein in CFA | |
| 500 μg | |
| Secondary | |
| 4°C | |
| Liquid |