missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, HRP, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen 31439

309.98 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.



Additional Details : Weight : 0.00150kg

Product Code. 10485715

  • € 329.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31439 has been successfully used in Western blot, and ICC applications. Antibody Specificity: This antibody reacts with heavy chains on mouse IgG but not with the light chains on most mouse immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins or mouse IgM. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine and horse serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.5 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA

Mouse IgG Fc Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG Fc Cross-Adsorbed in Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
Specifications

Specifications

Mouse IgG Fc Cross-Adsorbed
Polyclonal
HRP
Goat
1.5 mL
Secondary
4° C
IgG
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
0.8 mg/mL
PBS with 15mg/mL BSA and no preservative; pH 7.6
Antigen affinity chromatography
RUO
Mouse
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers