missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG Fc Secondary Antibody, DyLight 650, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60664C
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
| IgG Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat anti-Mouse IgG Fc fragment Secondary Antibody was produced by repeated immunization with Mouse IgG F(c) fragment in goat. | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
| DyLight 650 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction