missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG Fc Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75178
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
| IgG Fc | |
| Polyclonal | |
| FITC | |
| Goat | |
| Affinity purified | |
| RUO | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, Immunomicroscopy | |
| 1.0mg/mL | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Purified Mouse IgG, Fc fragment | |
| 2 mg | |
| Secondary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction