Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin-XX
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ B2763
Product Code. 10296492
  • € 228.00 / 1mg
Description
Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody for ChIP, ICC/IF, IHC, Western Blot
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
Biotin-XX
Goat
Purified
RUO
Mouse
Secondary Antibody
IgG
ChIP Assay, Immunohistochemistry, Western Blot, Immunocytochemistry
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains.
1 mg
Secondary
4°C