missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG/IgM (H+L) Secondary Antibody, Allophycocyanin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75217APC
This item is not returnable.
View return policy
Description
IgG/IgM (H+L) Polyclonal antibody specifically detects IgG/IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoassay
Specifications
| IgG/IgM (H+L) | |
| Polyclonal | |
| PBS | |
| Purified Mouse IgG + IgM | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoassay | |
| APC | |
| Goat | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction