missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG1 Heavy Chain Secondary Antibody, DyLight 594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7508DL594
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG1 Heavy Chain | |
| Polyclonal | |
| Western Blot : 1:1000 - 1:30000, ELISA : 1:1000- 1:30000, Immunohistochemistry : 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence : 1:200 - 1:2000, Immunohistochemistry-Paraffin : 1:200 - 1:2000 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50 mM Sodium Borate | |
| Mouse IgG1 | |
| 1 mL | |
| Immunology | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction