missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2a Heavy Chain Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7513V
This item is not returnable.
View return policy
Description
IgG2a Heavy Chain Polyclonal antibody specifically detects IgG2a Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG2a Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Mouse IgG2a | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction