missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2a Secondary Antibody, R-Phycoerythrin (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68511
This item is not returnable.
View return policy
Description
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Flow Cytometry
Specifications
| IgG2a | |
| Polyclonal | |
| R-PE | |
| PBS, 5% Sucrose | |
| IgG2a paraproteins from BALB/c mice | |
| 0.1 mg | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| 0.5 mg/mL | |
| Flow Cytometry 1:500 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction